Anti-Apg7 Rabbit Polyclonal Antibody

Toimittaja: Boster
BSBTPB9479EA 579 EUR
BSBTPB9479
Anti-Apg7 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for Ubiquitin-like modifier-activating enzyme ATG7(ATG7) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: Apg7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR