Anti-KIAA0652 Rabbit Polyclonal Antibody

Toimittaja: Boster
BSBTPB9480EA 579 EUR
BSBTPB9480
Anti-KIAA0652 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for Autophagy-related protein 13(ATG13) detection. Tested with WB in Human;Mouse.

Type: Primary
Antigen: KIAA0652
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR