Anti-ATP1A1 Rabbit Polyclonal Antibody

Toimittaja: Boster
BSBTPB9482EA 579 EUR
BSBTPB9482
Anti-ATP1A1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for Sodium/potassium-transporting ATPase subunit alpha-1(ATP1A1) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: ATP1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR