Anti-ICA1 Rabbit Polyclonal Antibody

Toimittaja: Boster
BSBTPB9495EA 579 EUR
BSBTPB9495
Anti-ICA1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for Islet cell autoantigen 1(ICA1) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: ICA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR