Anti-SOX5 Rabbit Polyclonal Antibody

Toimittaja: Boster
BSBTPB9507EA 579 EUR
BSBTPB9507
Anti-SOX5 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for Transcription factor SOX-5(SOX5) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: SOX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR