Anti-Ku70 Rabbit Polyclonal Antibody

Toimittaja: Boster
BSBTPB9520EA 550 EUR
BSBTPB9520
Anti-Ku70 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for X-ray repair cross-complementing protein 6(XRCC6) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: Ku70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR