Anti-APRIL Rabbit Polyclonal Antibody

Toimittaja: Boster
BSBTPB9523EA 0 EUR
BSBTPB9523
Anti-APRIL Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for Tumor necrosis factor ligand superfamily member 13 (TNFSF13) detection. Tested with WB, ELISA in Human.

Type: Primary
Antigen: APRIL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR