Anti-TARBP2 Antibody Pair

Toimittaja: Abnova
ABNOH00006895-PW3EA 835 EUR
ABNOH00006895-PW3
Anti-TARBP2 Antibody Pair
Vasta-aineet
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-TARBP2 (300 µg); 2. Antibody pair for Western Blot: Rabbit purified Polyclonal Anti-TARBP2 (50ug)

Type:
Antigen: TARBP2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR