Anti-TRIM55 Polyclonal Antibody Pair
Toimittaja: Abnova
ABNOH00084675-AP21EA 835 EUR
ABNOH00084675-AP21
Anti-TRIM55 Polyclonal Antibody Pair
Vasta-aineet
This Set comes with matched Antibody Pair to detect and quantify protein level of human TRIM55.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-TRIM55 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-TRIM55 (20 µg);
Type:
Antigen: TRIM55
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human