Anti-PDGFRA Antibody Pair

Toimittaja: Abnova
ABNODP0009EA 735 EUR
ABNODP0009
Anti-PDGFRA Antibody Pair
Vasta-aineet
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the PDGFRA protein, and the other against the specific Y574 phosphorylated site of PDGFRA for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-PDGFRA Y574 Rabbit Polyclonal Antibody (20 µl) With 0.09% sodium azide; 2. PDGFRA Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: PDGFRA
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR