Anti-SMAD3 Antibody Pair

Toimittaja: Abnova
ABNODP0012EA 884 EUR
ABNODP0012
Anti-SMAD3 Antibody Pair
Vasta-aineet
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the SMAD3 protein, and the other against the specific S208 phosphorylated site of SMAD3 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-SMAD3 S208 Rabbit Polyclonal Antibody (20 µl) In PBS (0.09% (w/v) sodium azide) 2. SMAD3 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: SMAD3
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR