Anti-NFKB1 Antibody Pair

Toimittaja: Abnova
ABNODP0021EA 884 EUR
ABNODP0021
Anti-NFKB1 Antibody Pair
Vasta-aineet
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the NFKB1 protein, and the other against the specific S337 phosphorylated site of NFKB1 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-NFKB1 S337 Rabbit Polyclonal Antibody (20 µl) With 0.09% sodium azide; 2. NFKB1 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: NFKB1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR