Anti-MCAM Polyclonal Antibody Pair

Toimittaja: Abnova
ABNODP0030EA 884 EUR
ABNODP0030
Anti-MCAM Polyclonal Antibody Pair
Vasta-aineet
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the MCAM protein, and the other against the specific Y641 phosphorylated site of MCAM for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-MCAM Y641 Rabbit Polyclonal Antibody (20 µl) In PBS (0.09% (w/v) sodium azide) 2. MCAM Mouse purified Polyclonal Antibody (40 µg) No additive; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: MCAM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR