Anti-IKBKB Antibody Pair

Toimittaja: Abnova
ABNODP0043EA 884 EUR
ABNODP0043
Anti-IKBKB Antibody Pair
Vasta-aineet
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the IKBKB protein, and the other against the specific Y199 phosphorylated site of IKBKB for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-IKBKB Y199 Rabbit Polyclonal Antibody (20 µl) With 0.09% sodium azide; 2. IKBKB Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: IKBKB
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR