Anti-ERBB2 Antibody Pair

Toimittaja: Abnova
ABNODP0044EA 884 EUR
ABNODP0044
Anti-ERBB2 Antibody Pair
Vasta-aineet
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the ERBB2 protein, and the other against the specific Y1221/Y1222 phosphorylated site of ERBB2 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-ERBB2 Y1221/Y1222 Rabbit Polyclonal Antibody (20 µl) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. ERBB2 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: ERBB2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR