Anti-BRAF Antibody Pair

Toimittaja: Abnova
ABNODP0052EA 884 EUR
ABNODP0052
Anti-BRAF Antibody Pair
Vasta-aineet
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the BRAF protein, and the other against the specific S445 phosphorylated site of BRAF for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-BRAF S445 Rabbit Polyclonal Antibody (20 µl) In PBS (0.09% (w/v) sodium azide) 2. BRAF Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: BRAF
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR