Anti-PRL + PRL Antibody Pair

Toimittaja: Abnova
ABNODP0054EA 735 EUR
ABNODP0054
Anti-PRL + PRL Antibody Pair
Vasta-aineet
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the PRL protein, and the other against the specific S163 phosphorylated site of PRL for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-PRL S163 Rabbit Polyclonal Antibody (20 µl) In PBS (0.09% (w/v) sodium azide) 2. PRL Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: PRL + PRL
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR