Anti-PDGFRB Antibody Pair

Toimittaja: Abnova
ABNODP0107EA 884 EUR
ABNODP0107
Anti-PDGFRB Antibody Pair
Vasta-aineet
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the PDGFRB protein, and the other against the specific Y857 phosphorylated site of PDGFRB for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-PDGFRB Y857 Rabbit Polyclonal Antibody (20 µl); 2. PDGFRB Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: PDGFRB
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR