Anti-CBL Polyclonal Antibody Pair

Toimittaja: Abnova
ABNOH00000867-AP21EA 884 EUR
ABNOH00000867-AP21
Anti-CBL Polyclonal Antibody Pair
Vasta-aineet
This Set comes with matched Antibody Pair to detect and quantify protein level of human CBL.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-CBL (100 µg); 2. Detection Antibody: Mouse Polyclonal Anti-CBL (40 µl);

Type:
Antigen: CBL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR