Anti-ELK1 Antibody Pair

Toimittaja: Abnova
ABNODP0146EA 884 EUR
ABNODP0146
Anti-ELK1 Antibody Pair
Vasta-aineet
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the ELK1 protein, and the other against the specific T417 phosphorylated site of ELK1 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-ELK1 T417 Rabbit Polyclonal Antibody (20 µl) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. ELK1 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: ELK1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR