Anti-AR Antibody Pair

Toimittaja: Abnova
ABNODP0169EA 884 EUR
ABNODP0169
Anti-AR Antibody Pair
Vasta-aineet
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the AR protein, and the other against the specific S213 phosphorylated site of AR for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-AR S213 Rabbit Polyclonal Antibody (20 µl) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. AR Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: AR
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR