Anti-KIT Antibody Pair

Toimittaja: Abnova
ABNODP0210EA 884 EUR
ABNODP0210
Anti-KIT Antibody Pair
Vasta-aineet
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the KIT protein, and the other against the specific Y721 phosphorylated site of KIT for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-KIT Y721 Rabbit Polyclonal Antibody (20 µl) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. KIT Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: KIT
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR