Anti-ERBB3 Antibody Pair

Toimittaja: Abnova
ABNODP0250EA 884 EUR
ABNODP0250
Anti-ERBB3 Antibody Pair
Vasta-aineet
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the ERBB3 protein, and the other against the specific Y1328 phosphorylated site of ERBB3 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-ERBB3 Y1328 Rabbit Polyclonal Antibody (20 µl) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. ERBB3 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: ERBB3
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR