Anti-VRK1 Antibody Pair

Toimittaja: Abnova
ABNOH00007443-AP51EA 835 EUR
ABNOH00007443-AP51
Anti-VRK1 Antibody Pair
Vasta-aineet
This Set comes with matched Antibody Pair to detect and quantify protein level of human VRK1.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-VRK1 (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-VRK1 (50 µg);

Type:
Antigen: VRK1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR