Anti-FGFR2 + PLCG1 Antibody Pair

Toimittaja: Abnova
ABNODI0421EA 794 EUR
ABNODI0421
Anti-FGFR2 + PLCG1 Antibody Pair
Vasta-aineet
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the FGFR2 protein, and the other against the PLCG1 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. FGFR2 Rabbit purified Polyclonal Antibody (20 µg) 2. PLCG1 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: FGFR2 + PLCG1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR