Anti-LILRA3 Antibody Pair
Toimittaja: Abnova
ABNOH00011026-AP51EA 835 EUR
ABNOH00011026-AP51
Anti-LILRA3 Antibody Pair
Vasta-aineet
This Set comes with matched Antibody Pair to detect and quantify protein level of human LILRA3.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-LILRA3 (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-LILRA3 (50 µg);
Type:
Antigen: LILRA3
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human