Anti-TCL1A Antibody Pair

Toimittaja: Abnova
ABNOH00008115-AP11EA 835 EUR
ABNOH00008115-AP11
Anti-TCL1A Antibody Pair
Vasta-aineet
This Set comes with matched Antibody Pair to detect and quantify protein level of human TCL1A.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-TCL1A (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-TCL1A, IgG1 kappa (20 µg);

Type:
Antigen: TCL1A
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR