Anti-SERPINA3 Polyclonal Antibody Pair
Toimittaja: Abnova
ABNOH00000012-PW1EA 884 EUR
ABNOH00000012-PW1
Anti-SERPINA3 Polyclonal Antibody Pair
Vasta-aineet
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-SERPINA3 (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-SERPINA3 (50 µg)
Type:
Antigen: SERPINA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human