Anti-GRN Antibody Pair

Toimittaja: Abnova
ABNOH00002896-AP51EA 835 EUR
ABNOH00002896-AP51
Anti-GRN Antibody Pair
Vasta-aineet
This Set comes with matched Antibody Pair to detect and quantify protein level of human GRN.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-GRN (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-GRN (50 µg);

Type:
Antigen: GRN
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR