Anti-OBFC1 Polyclonal Antibody Pair
Toimittaja: Abnova
ABNOH00079991-AP22EA 835 EUR
ABNOH00079991-AP22
Anti-OBFC1 Polyclonal Antibody Pair
Vasta-aineet
This Set comes with matched Antibody Pair to detect and quantify protein level of human OBFC1.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-OBFC1 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-OBFC1 (20 µg);
Type:
Antigen: OBFC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human