Anti-SEPT10 Polyclonal Antibody Pair

Toimittaja: Abnova
ABNOH00151011-PW1EA 835 EUR
ABNOH00151011-PW1
Anti-SEPT10 Polyclonal Antibody Pair
Vasta-aineet
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-SEPT10 (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-SEPT10 (50 µg)

Type:
Antigen: SEPT10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR