Anti-HMGB2 Mouse Monoclonal Antibody (Biotin)

Toimittaja: Abnova
ABNOH00003148-AP41EA 835 EUR
ABNOH00003148-AP41
Anti-HMGB2 Mouse Monoclonal Antibody (Biotin)
Vasta-aineet
This Set comes with matched Antibody Pair to detect and quantify protein level of human HMGB2.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-HMGB2 (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-HMGB2, IgG1 kappa (50 ?g);

Type:
Antigen: HMGB2
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR