Anti-KIT Mouse Monoclonal Antibody (Biotin)

Toimittaja: Abnova
ABNOH00003815-AP42EA 835 EUR
ABNOH00003815-AP42
Anti-KIT Mouse Monoclonal Antibody (Biotin)
Vasta-aineet
This Set comes with matched Antibody Pair to detect and quantify protein level of human KIT.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-KIT (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-KIT, IgG2b kappa (50 ?g);

Type:
Antigen: KIT
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR