Anti-CBL Mouse Monoclonal Antibody

Toimittaja: Abnova
ABNOH00000867-PW2EA 835 EUR
ABNOH00000867-PW2
Anti-CBL Mouse Monoclonal Antibody
Vasta-aineet
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-CBL (300 ?g); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-CBL (50 ?g)

Type:
Antigen: CBL
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR