Pressure lever cassettes
Toimittaja: Ismatec
ISMCIS3820AEA 165 EUR
ISMCIS3820A
Pressure lever cassettes
This optional pressure lever allows to set different tubing pressure for each channel. Depend on the tubing material and diameter an optimally adjusted tubing pressure can be set. To maintain constant flow rates it may be necessary to periodically adjust the tubing pressure.