Anti-BCHE Rabbit Polyclonal Antibody

Toimittaja: Boster
BSBTPB9526EA 579 EUR
BSBTPB9526
Anti-BCHE Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for Cholinesterase(BCHE) detection. Tested with WB in Mouse.

Type: Primary
Antigen: BCHE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR