Anti-CYP27B1 Rabbit Polyclonal Antibody

Toimittaja: Boster
BSBTPB9548EA 550 EUR
BSBTPB9548
Anti-CYP27B1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for 25-hydroxyvitamin D-1 alpha hydroxylase, mitochondrial (CYP27B1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: CYP27B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR