Laboratoriotakki, Howie style
Toimittaja: VWR Collection
113-8219EA 26.9 EUR
VWRI113-8219 VWRI113-8218 VWRI113-8215 VWRI113-8216 VWRI113-8220 VWRI113-8221
Laboratoriotakki, Howie style
These full-coverage lab coats made of 65% polyester and 35% cotton are treated with a dirt and water-repellent finish. They provide protection against dust, dirt and harmless liquid splashes. Ideal for use in non hazardous laboratory environments, in general hygienic areas and in hospitals and medical environments.
  • Mandarin collar, offset press-stud fastening
  • One mobile phone chest pocket with concealed press-stud fastening
  • Two hip patch pockets
  • Knitted cuffs
  • Washable at 60 °C
Colour: White
Length: 110 cm
Weight: 245 g/m²