Anti-G-CSF Antibody

Toimittaja: Boster
BSBTPB9577EA 579 EUR
BSBTPB9577
Anti-G-CSF Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for Granulocyte colony-stimulating factor(CSF3) detection. Tested with WB, ELISA in Human;Mouse.

Type: Primary
Antigen: G-CSF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR