Anti-Wnt3 Antibody

Toimittaja: Boster
BSBTPB9619EA 579 EUR
BSBTPB9619
Anti-Wnt3 Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for Proto-oncogene Wnt-3(WNT3) detection. Tested with WB in Human.

Type: Primary
Antigen: Wnt3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR