Goggles, uvex u-sonic
Toimittaja: Uvex
uvex ultrasonic uvex u-sonic
111-2275EA 29 EUR
111-2275 111-1706 111-1912 111-1705
Goggles, uvex u-sonic
Suojalasit, naamiomalliset
The ergonomic design with flexible soft component means the uvex u-sonic fits perfectly to every face its low weight and low pressure fit make them very comfortable to wear.
  • Combines hard and soft components which adapt to the wearer’s face creating soft, effective seal around the eyes
  • Fully adjustable, high quality head strap offers pressure-free fit
  • Prescription insert possible
  • Compact size and soft materials aid compatibility with respirators and other PPE items
  • Innovative ventilation system ensures pleasant comfortable interior environment
  • Large panorama lens is interchangeable
Can be used with add-on lenses. The integrated magnet technology makes it easy to swap out the add-on lens.
Weight: 69 g
Sertifikaatit: EN 166, EN 170, EN 172