Tubing, rubber, Versilon™ GS
Toimittaja: Saint-Gobain
VERN701590EA 742 EUR
VERN701590 VERN701550 VERN701520 VERN701510 VERN701620
Tubing, rubber, Versilon™ GS
Rubber, soft, beige.
Temperature resistance: −30 to +70 °C
Hardness (Shore durometer A): 42
Density: 1,07