Anti-LRP12 Rabbit Polyclonal Antibody

Toimittaja: Bioss
BOSSBS-6071REA 351 EUR
BOSSBS-6071R
Anti-LRP12 Rabbit Polyclonal Antibody
Vasta-aineet
Probable receptor, which may be involved in the internalization of lipophilic molecules and/or signal transduction. May act as a tumor suppressor.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: LRP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR