Anti-BPIL3 Rabbit Polyclonal Antibody

Toimittaja: Bioss
BOSSBS-8425REA 691 EUR
BOSSBS-8425R
Anti-BPIL3 Rabbit Polyclonal Antibody
Vasta-aineet
Members of the bactericidal/permeability-increasing protein family have antimicrobial properties and bind lipophilic substances, therefore targeting gram-negative bacteria. The bactericidal permeability increasing protein (BPI) is an antibacterial and endotoxin-neutralizing molecule that is abundant in the granules of polymorphonuclear leukocytes (neutrophil granules). Sharing structural and sequence homologies with BPI, BPIL3 (Bactericidal/permeability-increasing protein-like 3) is a 453 amino acid secreted protein that contains the family’s common conserved feature of two cysteine residues that are critical for protein function. While BPIL3 is primarily expressed at low levels in tonsils, it has been found to be upregulated in hypertrophic tonsil tissue, suggesting that it may play a role in the pathogenesis of inflamed disease tissue.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: BPIL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR