Anti-APLP2 Rabbit Polyclonal Antibody

Toimittaja: Bioss
BOSSBS-8462REA 351 EUR
BOSSBS-8462R
Anti-APLP2 Rabbit Polyclonal Antibody
Vasta-aineet
APLP2 is a human sperm membrane protein which contains a segment with high homology to the transmembrane-cytoplasmic domains of APP found in brain plaques of Alzheimer disease patients. The human amyloid precursor-like protein APLP2 is a highly conserved homolog of a sequence-specific DNA-binding mouse protein with an important function in the cell cycle. The gene which encodes APLP2 maps to human chromosome 11q24.May play a role in the regulation of hemostasis. The soluble form may have inhibitory properties towards coagulation factors. May interact with cellular G-protein signaling pathways. May bind to the DNA 5'-GTCACATG-3'(CDEI box). Inhibits trypsin, chymotrypsin, plasmin, factor XIA and plasma and glandular kallikrein.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: APLP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR