Human recombinant nesfatin 1
Toimittaja: Tonbo Biosciences
21-7114-M001EA 1090 EUR
TNBO21-7114-M001 TNBO21-7114-U020 TNBO21-7114-U100 TNBO21-7114-U500
Human recombinant nesfatin 1
Nesfatin-1 is a highly conserved metabolic polypetide derived through cleavage of its precursor protein, nucleobindin 2 (NUCB2). Nesfatin-1 is a secreted protein expressed in the hypothalamus and is implicated in regulation of appetite control and feeding behavior. It is described as an anorexigenic peptide and a satiety molecule. There is some indication that it plays a role in the adaptive response under stress conditions.