Human recombinant BAFF Receptor
Toimittaja: Tonbo Biosciences
21-7136-U500EA 897 EUR
TNBO21-7136-U500 TNBO21-7136-U010 TNBO21-7136-U050 TNBO21-7136-U100
Human recombinant BAFF Receptor
The B-cell activating factor (BAFF) receptor is a type III transmembrane protein belonging to the TNFR superfamily. It is expressed in spleen, lymph node and resting B-cells, with limited expression in activated B cells, resting CD4+ cells and peripheral blood leukocytes. BAFF/BAFFR activities are crucial to the survival and maturation of peripheral B cells. BAFF binding to BAFFR expressed on a subset of T cells has been shown to play a role in costimulating T cell activation.