Human recombinant BCMA
Toimittaja: Tonbo Biosciences
21-7137-U005EA 70.4 EUR
21-7137-U005 21-7137-U020 21-7137-U500 21-7137-U100
Human recombinant BCMA
Proteiinit
The full length B-Cell Maturation Antigen (BCMA) is a transmembrane protein consisting of 184 amino acids. It is a member of the TNF receptor superfamily and is preferentially expressed by mature B-cells. This protein may play a role in B- cell development and autoimmune response, and has been reported as essential for maintaining survival of plasma cells. It has been shown to bind to BAFF and APRIL, leading to NF-kappaB and MAPK8/JNK activation and stimulating IgM production.