Human recombinant IL36 beta
Toimittaja: Tonbo Biosciences
21-8372-U500EA 3140 EUR
TNBO21-8372-U500 TNBO21-8372-U100 TNBO21-8372-U010
Human recombinant IL36 beta
IL-36 beta, also known as IL-1F8, is a member of the IL-1 family of cytokines and binds to the IL-1R6 (IL-1Rrp2) receptor. It is reported to be expressed by resting and activated monocytes and B cells and at low levels in various tissues. It is highly expressed in psoriatic plaques. IL-36 beta displays proinflammatory effects and, like other IL-36 cytokines, can activate NF-kB and MAPK signaling pathways.