Anti-ZNF541 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP16706CEA 348 EUR
ABGEAP16706C
Anti-ZNF541 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ZNF541
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR